missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylbenzenesulfonic acid, 99%, Thermo Scientific Chemicals

Product Code. 11450398
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-44-8
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00007908
InChI Key LTPSRQRIPCVMKQ-UHFFFAOYSA-N
Synonym 4-aminotoluene-3-sulfonic acid, ptmsptmsa, ptms, ptmsa, benzenesulfonic acid, 2-amino-5-methyl, 6-amino-m-toluenesulfonic acid, red 4b acid, p-toluidine-2-sulfonic acid, p-toluidine-m-sulfonic acid, m-toluenesulfonic acid, 6-amino
PubChem CID 6934
IUPAC Name 2-amino-5-methylbenzenesulfonic acid
SMILES CC1=CC=C(N)C(=C1)S(O)(=O)=O

Specifications

Quantity 50 g
UN Number UN3261
Beilstein 2211509
Formula Weight 187.22
Percent Purity 99%
Chemical Name or Material 2-Amino-5-methylbenzenesulfonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.