missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzonitrile, 95%, Thermo Scientific Chemicals

Product Code. 11477096
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17420-30-3
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00007362
InChI Key MGCGMYPNXAFGFA-UHFFFAOYSA-N
Synonym 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j
PubChem CID 28532
IUPAC Name 2-amino-5-nitrobenzonitrile
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C#N)N

Specifications

Melting Point 207°C to 210°C
Quantity 100 g
Formula Weight 163.14
Percent Purity 95%
Chemical Name or Material 2-Amino-5-nitrobenzonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'