missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals

Product Code. 10552294
Click to view available options
Quantity:
25 g
50 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Brown to Brown-Rust
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether
Formula Weight 154.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrophenol, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-5-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.