missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-trifluoromethyl-1,3,4-thiadiazole, 98%, Thermo Scientific Chemicals

Product Code. 11408926
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10444-89-0
Molecular Formula C3H2F3N3S
Molecular Weight (g/mol) 169.125
MDL Number MFCD00003109
InChI Key LTEUXHSAYOSFGQ-UHFFFAOYSA-N
Synonym 5-trifluoromethyl-1,3,4-thiadiazol-2-amine, 2-amino-5-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine, 5-trifluoromethyl, 5-trifluoromethyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-trifluoromethyl, 5-trifluoromethyl-1,3,4-thiadiazole-2-ylamine, zlchem 1059, pubchem14503, maybridge1_003448
PubChem CID 82634
IUPAC Name 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
SMILES C1(=NN=C(S1)N)C(F)(F)F

Specifications

Melting Point 222°C to 227°C
Quantity 1 g
Beilstein 513140
Formula Weight 169.13
Percent Purity 98%
Chemical Name or Material 2-Amino-5-trifluoromethyl-1,3,4-thiadiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'