missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromobenzothiazole, 95%, Thermo Scientific Chemicals

115.00 € - 511.00 €

Chemical Identifiers

CAS 15864-32-1
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.10
MDL Number MFCD00152229
InChI Key VZEBSJIOUMDNLY-UHFFFAOYSA-N
Synonym 2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
PubChem CID 85149
IUPAC Name 6-bromo-1,3-benzothiazol-2-amine
SMILES NC1=NC2=CC=C(Br)C=C2S1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11986791
View Documents
Thermo Scientific Acros
442290050
5 g
115.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11996791
View Documents
Thermo Scientific Acros
442290250
25 g
511.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15864-32-1
229.10
VZEBSJIOUMDNLY-UHFFFAOYSA-N
85149
NC1=NC2=CC=C(Br)C=C2S1
C7H5BrN2S
MFCD00152229
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
6-bromo-1,3-benzothiazol-2-amine

Specifications

15864-32-1
100.0
White
94% min. (HPLC)
C7H5BrN2S
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
NC1=NC2=CC=C(Br)C=C2S1
229.10
229.1
Powder
94.0
212°C to 214°C
Authentic
Glass bottle
MFCD00152229
VZEBSJIOUMDNLY-UHFFFAOYSA-N
6-bromo-1,3-benzothiazol-2-amine
85149
95%
2-Amino-6-bromobenzothiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Contaminated work clothing should not be allowed out of the workplace.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 2-Amino-6-bromobenzothiazole

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only