missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Product Code. 15441919
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27048-04-0
Molecular Formula C5H4ClN3O2
Molecular Weight (g/mol) 173.56
MDL Number MFCD00044350
InChI Key WERABQRUGJIMKQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloro-3-nitropyridine, 2-amino-3-nitro-6-chloropyridine, 6-chloro-3-nitro-2-pyridylamine, 6-chloro-3-nitro-2-pyridinamine, 6-amino-2-chloro-5-nitropyridine, 6-chloro-3-nitropyridin-2-ylamine, 6-chloro-3-nitro-pyridin-2-ylamine, 2-pyridinamine, 6-chloro-3-nitro, 6-chloro-3-nitro-pyridin-2-amine, zlchem 113
PubChem CID 2724035
IUPAC Name 6-chloro-3-nitropyridin-2-amine
SMILES NC1=NC(Cl)=CC=C1[N+]([O-])=O

Specifications

Melting Point 192°C to 196°C
Color Yellow
Quantity 25 g
UN Number UN2811
Beilstein 474607
Solubility Information Slightly soluble in water. Soluble in dimethyl sulfoxide and chloroform.
Formula Weight 173.56
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2-Amino-6-chloro-3-nitropyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.