missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 10148043
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-76-4
Molecular Formula C7H5FNO2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00067781
InChI Key RWSFZKWMVWPDGZ-UHFFFAOYSA-M
Synonym 6-fluoroanthranilic acid, anthralic acid, 6-fluoro, benzoic acid, 2-amino-6-fluoro, 6-amino-2-fluorobenzoic acid, 2-amino-6-fluoro-benzoic acid, 2,amino-6-fluorobenzoic acid, 4owo, pubchem1324, 6-fluoroanthanilic acid, acmc-209jun
PubChem CID 521142
SMILES NC1=CC=CC(F)=C1C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 170°C to 172°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 5 g
IUPAC Name 2-amino-6-fluorobenzoate
Formula Weight 155.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-fluorobenzoic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'