missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-purinethiol, 95%, ACROS Organics™

Product Code. 10771345
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >360.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 250 mg
Beilstein 26, IV, 3926
Merck Index 15, 9490
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in an aqueous naoh solution
Formula Weight 167.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Amino-6-purinethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-6-purinethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.