missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoacetophenone hydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11422240
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5468-37-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00012873
InChI Key CVXGFPPAIUELDV-UHFFFAOYSA-N
Synonym 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
PubChem CID 2723597
IUPAC Name 2-amino-1-phenylethanone;hydrochloride
SMILES Cl.NCC(=O)C1=CC=CC=C1

Specifications

Melting Point ∽194°C (decomposition)
Odor Odorless
Quantity 50 g
Formula Weight 171.63
Percent Purity 98%
Chemical Name or Material 2-Aminoacetophenone hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'