missing translation for 'onlineSavingsMsg'
Learn More
2-Aminoanthracene, 94%, Thermo Scientific Chemicals Product Code.: 11389075

2-Aminoanthracene, 94%, Thermo Scientific Chemicals

Product Code. 11389075
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11389075

Brand: Thermo Scientific Alfa Aesar L14269.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 613-13-8
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00003582
InChI Key YCSBALJAGZKWFF-UHFFFAOYSA-N
Synonym 2-aminoanthracene, 2-anthramine, 2-anthracenamine, 2-anthrylamine, 2-anthracylamine, beta-aminoanthracene, 2-anthracenamide, ccris 22, .beta.-aminoanthracene, dsstox_cid_4458
PubChem CID 11937
ChEBI CHEBI:34260
IUPAC Name anthracen-2-amine
SMILES C1=CC=C2C=C3C=C(C=CC3=CC2=C1)N

Specifications

Melting Point 235°C to 239°C
Quantity 5 g
Beilstein 2209414
Formula Weight 193.25
Percent Purity 94%
Chemical Name or Material 2-Aminoanthracene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminoanthracene, 94%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.