missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenesulfonamide, 98%, Thermo Scientific Chemicals

Product Code. 11494987
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3306-62-5
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.202
MDL Number MFCD00007932
InChI Key YAZSBRQTAHVVGE-UHFFFAOYSA-N
Synonym orthanilamide, o-aminobenzenesulfonamide, o-sulfanilamide, benzenesulfonamide, 2-amino, 2-amino-benzenesulfonamide, benzenesulfonamide, o-amino, 2-aminobenzene-1-sulfonamide, o-aminobenzenesulphonamide, chembl6705, benzenesulfonamide, 2-amino-9ci
PubChem CID 72894
IUPAC Name 2-aminobenzenesulfonamide
SMILES C1=CC=C(C(=C1)N)S(=O)(=O)N

Specifications

Melting Point 153°C to 157°C
Odor Odorless
Quantity 25 g
Formula Weight 172.21
Percent Purity 98%
Chemical Name or Material 2-Aminobenzenesulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'