missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Aminobutane, 98%, Thermo Scientific Chemicals

Product Code. 11449923
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

(±)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive air and light. Incompatible with strong oxidizing agents. Incompatible with strong oxidizing agents, strong acids and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13952-84-6
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
MDL Number MFCD00008094
InChI Key BHRZNVHARXXAHW-UHFFFAOYNA-N
Synonym sec-butylamine, 2-butanamine, 2-aminobutane, 2-butylamine, 1-methylpropylamine, butafume, tutane, 1-methylpropanamine, deccotane, frucote
PubChem CID 24874
ChEBI CHEBI:74526
IUPAC Name butan-2-amine
SMILES CCC(C)N

Specifications

Melting Point -104°C
Density 0.722
pH 12.5
Boiling Point 63°C to 64°C
Flash Point −19°C (−2°F)
Odor Amine-like
Refractive Index 1.393
Quantity 500 mL
UN Number UN2733
Beilstein 1361345
Sensitivity Air Sensitive
Merck Index 14,1544
Solubility Information Soluble in water.
Formula Weight 73.14
Percent Purity 98%
Chemical Name or Material (+/-)-2-Aminobutane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'