Learn More
2-Aminoethanethiol, 95%, Thermo Scientific Chemicals
77.30 € - 211.00 €
Chemical Identifiers
CAS | 60-23-1 |
---|---|
Molecular Formula | C2H7NS |
Molecular Weight (g/mol) | 77.15 |
MDL Number | MFCD00008196 |
InChI Key | UFULAYFCSOUIOV-UHFFFAOYSA-N |
Synonym | cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten |
PubChem CID | 6058 |
ChEBI | CHEBI:17141 |
IUPAC Name | 2-aminoethanethiol |
SMILES | NCCS |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11331143
|
Thermo Scientific Acros
434710050 |
5 g | Glass bottle |
77.30 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955751
|
Thermo Scientific Acros
434710250 |
25 g | Glass bottle |
211.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
60-23-1 | |
77.15 | |
UFULAYFCSOUIOV-UHFFFAOYSA-N | |
6058 | |
2-aminoethanethiol |
C2H7NS | |
MFCD00008196 | |
cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten | |
CHEBI:17141 | |
NCCS |
Spezifikation
60-23-1 | |
100.0 | |
White to Yellow | |
94% min. (Iodimetry) | |
NH2CH2CH2SH | |
15,2776 | |
Solubility in water: freely soluble. | |
NCCS | |
77.15 | |
CHEBI:17141 | |
95% | |
2-Aminoethanethiol |
94.0 | |
95.0°C | |
Authentic | |
C2H7NS | |
MFCD00008196 | |
cysteamine, mercaptamine, thioethanolamine, becaptan, mercamine, cysteinamine, beta-mercaptoethylamine, 2-mercaptoethylamine, cysteamin, lambraten | |
UFULAYFCSOUIOV-UHFFFAOYSA-N | |
2-aminoethanethiol | |
6058 | |
77.15 | |
Powder |
Sicherheit und Handhabung
GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-463-
- 2-Aminoethanethiol
Signalwort
- Warning
Gefahrenkategorie
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Sicherheitshinweis
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only