missing translation for 'onlineSavingsMsg'
Learn More
2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals Product Code.: 11497947

2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals

Product Code. 11497947
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11497947

Brand: Thermo Scientific Alfa Aesar A16606.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminoethyl diphenylborinate is used for the densitometric determination of flavonoids and TRP activator. It is also used to manipulate intracellular release of calcium ions and in preparation of apoptotic inducers. It serves as a reactant for preparation of apoptotic inducers and in membrane-permeable SOCE (store operated calcuym entry) inhibitor.

Solubility
Soluble in ethanol and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 524-95-8
Molecular Formula C14H16BNO
Molecular Weight (g/mol) 225.10
MDL Number MFCD00014823
InChI Key BLZVCIGGICSWIG-UHFFFAOYSA-N
Synonym 2-aminoethyl diphenylborinate, 2-apb, 2-aminoethoxy diphenylborane, 2-aminoethoxydiphenylborane, 2-aminoethoxydiphenylborate, 2-aminoethoxydiphenyl borate, o-2-aminoethyl diphenylborinic acid, b-2-aminoethoxy diphenylborane, 2-diphenylboryl oxy ethanamine, borinic acid, o-2-aminoethyl diphenylShow MoreShow Less
PubChem CID 1598
SMILES NCCOB(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 187°C to 190°C
Quantity 25 g
Beilstein 2942273
Solubility Information Soluble in ethanol and methanol.
IUPAC Name 2-diphenylboranyloxyethanamine
Formula Weight 225.1
Percent Purity 98%
Chemical Name or Material 2-Aminoethyl diphenylborinate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254765 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog NumberA16606.06,A16606.14
Lot Number10244943 Certificate TypeCertificate of Analysis Date06/12/2022 Catalog NumberA16606.06,A16606.14
Lot Number10240570 Certificate TypeCertificate of Analysis Date21/05/2022 Catalog NumberA16606.06,A16606.14
Lot Number10223549 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16606.06,A16606.14
Lot Number10207684 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA16606.06,A16606.14
compliance-icons
Product Identifier
  • 2-Aminoethyl diphenylborinate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : (2-Aminoethoxy)(diphenyl)borane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminoethyl diphenylborinate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.