missing translation for 'onlineSavingsMsg'
Learn More

2-Aminopyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15471196
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Aminopyrimidine-5-boronic acid pinacol ester is used in the preparation of phosphoinositide-3-kinases (PI3K) inhibitors.

Solubility
Slightly soluble in water. Soluble in chloroform.

Notes
Incompatible with strong oxidizing agents, reducing agents and acids. Store at 4-2°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402960-38-7
Molecular Formula C10H16BN3O2
Molecular Weight (g/mol) 221.067
MDL Number MFCD06795685
InChI Key BPQVMIDUTRJYSC-UHFFFAOYSA-N
Synonym 2-aminopyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-aminopyrimidine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrimidin-2-amine, 2-aminopyrimidin-5-yl boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinylamine, 2-2-aminopyrimidin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 17750212
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)N

Specifications

Melting Point 206°C to 208°C
Quantity 1 g
Solubility Information Slightly soluble in water. Soluble in chloroform.
Formula Weight 221.06
Percent Purity 96%
Chemical Name or Material 2-Aminopyrimidine-5-boronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'