missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminopyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2-Aminopyrimidine-5-boronic acid pinacol ester is used in the preparation of phosphoinositide-3-kinases (PI3K) inhibitors.
Solubility
Slightly soluble in water. Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and acids. Store at 4-2°C.
2-Aminopyrimidine-5-boronic acid pinacol ester is used in the preparation of phosphoinositide-3-kinases (PI3K) inhibitors.
Solubility
Slightly soluble in water. Soluble in chloroform.
Notes
Incompatible with strong oxidizing agents, reducing agents and acids. Store at 4-2°C.
Specifications
Specifications
Melting Point | 206°C to 208°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. Soluble in chloroform. |
Formula Weight | 221.06 |
Percent Purity | 96% |
Chemical Name or Material | 2-Aminopyrimidine-5-boronic acid pinacol ester |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction