Learn More
DiscoverX 2-Arachidonylglycerol
2-Arachidonylglycerol is an agonist for GPR55, CB1 and CB2. Provided as 10mg of dry powder.
Brand:  DiscoverX 92-1226
Encompass
Product Code. 17302549
  • 658.50 € / 10mg
Explore more special offers
Description
Description
2-Arachidonylglycerol is an endocannabinoid ester formed by arachidonic acid and glycerol that is expressed in the CNS at much higher levels than anandamide. Has neuromodulatory effects, and is a potent agonist of the GPR55, CB1, and CB2 receptors. Provided as 10mg of dry powder.