missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyl-N-Boc-D-proline, 95%, Thermo Scientific™

Product Code. 15488747
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 706806-61-3
Molecular Formula C17H23NO4
Molecular Weight (g/mol) 305.374
MDL Number MFCD06659153
InChI Key JUUNPRGYFCIXSM-KRWDZBQOSA-N
Synonym s-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-s-alpha-benzyl-proline, boc-s-a-benzylproline, s-2-benzyl-1-boc-2-pyrrolidinecarboxylic acid, 2s-2-benzyl-1-tert-butoxycarbonyl pyrrolidine-2-carboxylic acid, boc-, a-benzyl-d-proline, 2-benzyl-n-boc-d-proline, boc-s-, a-benzyl-pro-oh, boc-s-alpha-benzyl-pro-oh, 2-benzyl-1-tert-butoxycarbonyl-d-proline
PubChem CID 6950294
IUPAC Name (2S)-2-benzyl-1-[(2-methylpropan-2-yl)oxycarbonyl]pyrrolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCCC1(CC2=CC=CC=C2)C(=O)O

Specifications

Melting Point 146°C
Quantity 250 mg
Formula Weight 305.37
Percent Purity 95%
Chemical Name or Material 2-Benzyl-N-Boc-D-proline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'