Learn More
2-Benzylaminopyridine, 98%, Thermo Scientific™
28.30 € - 290.00 €
Chemical Identifiers
CAS | 6935-27-9 |
---|---|
Molecular Formula | C12H12N2 |
Molecular Weight (g/mol) | 184.242 |
MDL Number | MFCD00006251 |
InChI Key | WYHXNQXDQQMTQI-UHFFFAOYSA-N |
Synonym | 2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g |
PubChem CID | 23362 |
IUPAC Name | N-benzylpyridin-2-amine |
SMILES | C1=CC=C(C=C1)CNC2=CC=CC=N2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460587
|
Thermo Scientific Alfa Aesar
B22441.18 |
50 g |
28.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470587
|
Thermo Scientific Alfa Aesar
B22441.30 |
250 g | N/A | ||||||
11450587
|
Thermo Scientific Alfa Aesar
B22441.0B |
1000 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
6935-27-9 | |
184.242 | |
WYHXNQXDQQMTQI-UHFFFAOYSA-N | |
23362 | |
C1=CC=C(C=C1)CNC2=CC=CC=N2 |
C12H12N2 | |
MFCD00006251 | |
2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g | |
N-benzylpyridin-2-amine |
Specifications
6935-27-9 | |
182°C (12 mmHg) | |
C12H12N2 | |
134309 | |
WYHXNQXDQQMTQI-UHFFFAOYSA-N | |
N-benzylpyridin-2-amine | |
23362 | |
98% |
93°C to 97°C | |
153°C (307°F) | |
MFCD00006251 | |
2-benzylaminopyridine, 2-pyridinamine, n-phenylmethyl, 2-benzylamino pyridine, pyridine, 2-benzylamino, n-benzyl-2-pyridinamine, benzyl-2-pyridylamine, enamine_005854, cbmicro_010916, n-2-pyridyl benzylamine, acmc-209o7g | |
C1=CC=C(C=C1)CNC2=CC=CC=N2 | |
184.242 | |
184.24 | |
2-Benzylaminopyridine |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 230-060-5
missing translation for 'rtecsNumber' : US2700000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures