missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-BFI hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity I2 ligand (Ki = 9.8 nM). Putative I2 agonist; potentiates morphine antinociception. Produces ipsiversive rotational behavior in rats with a full 6-OHDA lesion of the nigrostriatal tract.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 222.67 |
Percent Purity | >98% |
Chemical Name or Material | 2-BFI hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'