missing translation for 'onlineSavingsMsg'
Learn More
2-(Boc-amino)-4-methylpyridine, 97%, Thermo Scientific™ Product Code.: 15487587

2-(Boc-amino)-4-methylpyridine, 97%, Thermo Scientific™

Product Code. 15487587
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15487587

Brand: Thermo Scientific Alfa Aesar H35892.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90101-20-5
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD07776933
InChI Key SABMAMDNSRZJBK-UHFFFAOYSA-N
Synonym 4-methyl-pyridin-2-yl-carbamic acid tert-butyl ester, tert-butyl 4-methylpyridin-2-yl carbamate, tert-butyl n-4-methylpyridin-2-yl carbamate, 2-boc-amino-4-methylpyridine, tert-butyl 4-methylpyridin-2-ylcarbamate, 4-methylpyridin-2-yl carbamic acid tert-butyl ester, carbamic acid, 4-methyl-2-pyridinyl-, 1,1-dimethylethyl ester, ambkkkkk140, tert-butyl 4-methyl-2-pyridinylcarbamate
PubChem CID 11063648
IUPAC Name tert-butyl N-(4-methylpyridin-2-yl)carbamate
SMILES CC1=CC(=NC=C1)NC(=O)OC(C)(C)C

Specifications

Melting Point 118°C to 119°C
Quantity 250 mg
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)-4-methylpyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Boc-amino)-4-methylpyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.