missing translation for 'onlineSavingsMsg'
Learn More

2-(Boc-amino)-5-cyanopyridine, 97%, Thermo Scientific™

Product Code. 11474490
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902837-44-9
Molecular Formula C11H13N3O2
Molecular Weight (g/mol) 219.24
MDL Number MFCD07781163
InChI Key JDQQNZYKHZVLAM-UHFFFAOYSA-N
Synonym 2-boc-amino-5-cyanopyridine, tert-butyl 5-cyanopyridin-2-yl carbamate, tert-butyl 5-cyanopyridin-2-ylcarbamate, tert-butyl n-5-cyanopyridin-2-yl carbamate, 6-boc-amino nicotinonitrile, 6-boc-amino pyridine-3-carbonitrile, tert-butyl 5-cyano-2-pyridinylcarbamate, 2-boc-amino pyridine-5-carbonitrile, tert-butoxy-n-5-cyano 2-pyridyl carboxamide, 5-cyano-pyridin-2-yl-carbamic acid tert-butyl ester
PubChem CID 42553119
IUPAC Name tert-butyl N-(5-cyanopyridin-2-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=NC=C(C=C1)C#N

Specifications

Melting Point 170°C to 175°C
Quantity 1 g
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)-5-cyanopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'