missing translation for 'onlineSavingsMsg'
Learn More
2-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals Product Code.: 11442210

2-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11442210
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11442210

Brand: Thermo Scientific Alfa Aesar H29020.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 159624-15-4
Molecular Formula C17H26BNO4
Molecular Weight (g/mol) 319.208
MDL Number MFCD03411943
InChI Key LVHGGVGVAUJQBB-UHFFFAOYSA-N
Synonym tert-butyl 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-amino benzeneboronic acid pinacol ester, 2-n-boc-amino phenylboronic acid pinacol ester, tert-butyl-n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, tert-butyl n-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenylboronic acid pinacol ester, tert-butyl n-2-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl carbamate, 2-boc-aminophenyl boronic acid, pinacol ester, 2-boc-amino phenylboronic acid pinacol ester, 2-tert-butoxycarbonylamino phenylboronic acid, pinacol ester
PubChem CID 2773231
IUPAC Name tert-butyl N-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]carbamate
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2NC(=O)OC(C)(C)C

Specifications

Melting Point 98°C to 100°C
Quantity 5 g
Formula Weight 319.21
Percent Purity 97%
Chemical Name or Material 2-(Boc-amino)benzeneboronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10151859 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog NumberH29020.06,H29020.03
Lot Number10151859 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29020.06,H29020.03
Lot Number10144603 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH29020.06,H29020.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Boc-amino)benzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.