missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™ Product Code.: 10471304

2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™

Product Code. 10471304
25 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10471304

Brand: Thermo Scientific Maybridge CC04005EB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17570-98-8
Molecular Formula C7H7Br2NO
Molecular Weight (g/mol) 280.947
InChI Key BYKVUGZUYJUSKD-UHFFFAOYSA-N
Synonym 2-bromoacetyl pyridine hydrobromide, 2-bromo-1-pyridin-2-yl ethanone hydrobromide, 2-bromo-1-2-pyridinyl-1-ethanone hydrobromide, 2-2-bromoacetyl pyridine hydrobromide, 2-bromoacetylpyridine hydrobromide, 2-bromo-1-pyridin-2-ylethanone hydrobromide, 2-bromo-1-2-pyridinyl ethanone hydrobromide, 2-bromo-1-pyridin-2-yl ethan-1-one hydrobromide, zlchem 1071, pubchem21374
PubChem CID 2776231
IUPAC Name 2-bromo-1-pyridin-2-ylethanone;hydrobromide
SMILES C1=CC=NC(=C1)C(=O)CBr.Br

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 280.95
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-1-(2-pyridinyl)-1-ethanone hydrobromide, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.