missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[3-(4-chlorophenyl)-5-isoxazolyl]-1-ethanone, 97%, Thermo Scientific™

14.50 € - 118.00 €

Chemical Identifiers

CAS 258506-49-9
Molecular Formula C11H7BrClNO2
Molecular Weight (g/mol) 300.536
MDL Number MFCD00662759
InChI Key OUHGJKFYGYXITB-UHFFFAOYSA-N
Synonym 2-bromo-1-3-4-chlorophenyl-5-isoxazolyl-1-ethanone, 5-bromoacetyl-3-4-chlorophenyl isoxazole, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl-isoxazol-5-yl ethanone, 2-bromo-1-3-4-chloro-phenyl-isoxazol-5-yl-ethanone, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethan-1-one, pubchem8718
PubChem CID 2799661
IUPAC Name 2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethanone
SMILES C1=CC(=CC=C1C2=NOC(=C2)C(=O)CBr)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10223142
SDS View Documents
Thermo Scientific Maybridge
SPB05675CB
250 mg
14.50 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10170702
SDS View Documents
Thermo Scientific Maybridge
SPB05675DA
1 g
37.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10301645
SDS View Documents
Thermo Scientific Maybridge
SPB05675DE
5 g
118.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

258506-49-9
300.536
OUHGJKFYGYXITB-UHFFFAOYSA-N
2799661
C1=CC(=CC=C1C2=NOC(=C2)C(=O)CBr)Cl
C11H7BrClNO2
MFCD00662759
2-bromo-1-3-4-chlorophenyl-5-isoxazolyl-1-ethanone, 5-bromoacetyl-3-4-chlorophenyl isoxazole, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl-isoxazol-5-yl ethanone, 2-bromo-1-3-4-chloro-phenyl-isoxazol-5-yl-ethanone, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethan-1-one, pubchem8718
2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethanone

Specifications

258506-49-9
97%
C11H7BrClNO2
2-bromo-1-3-4-chlorophenyl-5-isoxazolyl-1-ethanone, 5-bromoacetyl-3-4-chlorophenyl isoxazole, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethan-1-one, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl isoxazol-5-yl ethanone, 2-bromo-1-3-4-chlorophenyl-isoxazol-5-yl ethanone, 2-bromo-1-3-4-chloro-phenyl-isoxazol-5-yl-ethanone, 2-bromo-1-3-4-chlorophenyl-1,2-oxazol-5-yl ethan-1-one, pubchem8718
C1=CC(=CC=C1C2=NOC(=C2)C(=O)CBr)Cl
300.536
300.54
2-Bromo-1-[3-(4-chlorophenyl)-5-isoxazolyl]-1-ethanone
97.0
Amber Glass Bottle
MFCD00662759
OUHGJKFYGYXITB-UHFFFAOYSA-N
2-bromo-1-[3-(4-chlorophenyl)-1,2-oxazol-5-yl]ethanone
2799661
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.