missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3-benzothiazole, 95%, Thermo Scientific™

Product Code. p-2697879
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10475502

Brand: Thermo Scientific Maybridge CC06010CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2516-40-7
Molecular Formula C7H4BrNS
Molecular Weight (g/mol) 214.08
MDL Number MFCD02681887
InChI Key DRLMMVPCYXFPEP-UHFFFAOYSA-N
Synonym 2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
PubChem CID 612040
IUPAC Name 2-bromo-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 214.09
Percent Purity 97%
Chemical Name or Material 2-Bromo-1, 3-benzothiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.