missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one 95%, Thermo Scientific™

Product Code. 10062264
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14731-14-7
Molecular Formula C11H8BrNO2
Molecular Weight (g/mol) 266.09
MDL Number MFCD00173898
InChI Key XTBXGZOVSCTNEC-UHFFFAOYSA-N
Synonym 5-bromoacetyl-3-phenylisoxazole, 2-bromo-1-3-phenylisoxazol-5-yl ethan-1-one, 2-bromo-1-3-phenylisoxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethanone, 2-bromo-1-3-phenyl-1,2-oxazol-5-yl ethan-1-one, 2-bromo-1-3-phenyl-isoxazol-5-yl-ethanone, ethanone, 2-bromo-1-3-phenyl-5-isoxazolyl, pubchem8717, buttpark 43\57-72
PubChem CID 2735532
IUPAC Name 2-bromo-1-(3-phenyl-1,2-oxazol-5-yl)ethanone
SMILES BrCC(=O)C1=CC(=NO1)C1=CC=CC=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 266.09
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(3-phenylisoxazol-5-yl)ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.