missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(2-thienyl)phenyl]-1-ethanone, ≥97%, Thermo Scientific™

35.40 € - 285.00 €

Chemical Identifiers

CAS 128746-80-5
Molecular Formula C12H9BrOS
Molecular Weight (g/mol) 281.167
MDL Number MFCD06659059
InChI Key QFHKNYPNLSJEQI-UHFFFAOYSA-N
PubChem CID 2795250
IUPAC Name 2-bromo-1-(4-thiophen-2-ylphenyl)ethanone
SMILES C1=CSC(=C1)C2=CC=C(C=C2)C(=O)CBr
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10681075
View Documents Promotion Details
Thermo Scientific Maybridge
CC12005CB
250 mg
35.40 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10113942
View Documents Promotion Details
Thermo Scientific Maybridge
CC12005DA
1 g
93.40 €
1g
Please to purchase this item. Need a web account? Register with us today!
10102884
View Documents Promotion Details
Thermo Scientific Maybridge
CC12005DE
5 g
285.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

128746-80-5
281.167
QFHKNYPNLSJEQI-UHFFFAOYSA-N
2-bromo-1-(4-thiophen-2-ylphenyl)ethanone
C12H9BrOS
MFCD06659059
2795250
C1=CSC(=C1)C2=CC=C(C=C2)C(=O)CBr

Specifications

128746-80-5
100.0
Amber glass bottle
MFCD06659059
QFHKNYPNLSJEQI-UHFFFAOYSA-N
2-bromo-1-(4-thiophen-2-ylphenyl)ethanone
2795250
97%
96.5
95%
C12H9BrOS
2-bromo-1-4-2-thienyl phenyl-1-ethanone, ethanone,2-bromo-1-4-2-thienyl phenyl, 2-bromo-1-4-thiophen-2-yl phenyl ethanone, acmc-20msyh, 2-bromo-1-thien-2yphenyl ethanone, 2-bromo-4'-thien-2-yl acetophenone, 2-bromo-1-4-thiophen-2-ylphenyl ethanone, ethanone, 2-bromo-1-4-2-thienyl phenyl, 2-bromo-1-4-thien-2-yl phenyl ethan-1-one
C1=CSC(=C1)C2=CC=C(C=C2)C(=O)CBr
281.167
281.17
2-Bromo-1-[4-(2-thienyl)phenyl]-1-ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

SDS
Documents

Documents

Product Certifications