missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-(4-pentylphenyl)ethan-1-one, 95%, Thermo Scientific™

Product Code. 10180622
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 64328-68-3
Molecular Formula C13H17BrO
Molecular Weight (g/mol) 269.182
MDL Number MFCD00218837
InChI Key AVYNDJWQMUOSJZ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
PubChem CID 2775670
IUPAC Name 2-bromo-1-(4-pentylphenyl)ethanone
SMILES CCCCCC1=CC=C(C=C1)C(=O)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula CH3(CH2)4C6H4C(O)CH2Br
Quantity 1 g
Formula Weight 269.18
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-pentylphenyl)ethan-1-one
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'