missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one, Tech., Thermo Scientific™

11.30 € - 29.00 €

Chemical Identifiers

CAS 383-53-9
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.05
MDL Number MFCD00126489
InChI Key HEMROKPXTCOASZ-UHFFFAOYSA-N
Synonym 2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
PubChem CID 321979
IUPAC Name 2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
SMILES FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10752133
SDS View Documents
Thermo Scientific Maybridge
SPB06282DA
1 g Amber Glass Bottle
11.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
10009461
SDS View Documents
Thermo Scientific Maybridge
SPB06282EA
10 g Amber Glass Bottle
29.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

383-53-9
267.05
HEMROKPXTCOASZ-UHFFFAOYSA-N
321979
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
C9H6BrF3O
MFCD00126489
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone

Specifications

383-53-9
C9H6BrF3O
MFCD00126489
HEMROKPXTCOASZ-UHFFFAOYSA-N
2-bromo-1-[4-(trifluoromethyl)phenyl]ethanone
321979
Tech.
2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one
Tech.
CF3C6H4C(O)CH2Br
2-bromo-1-4-trifluoromethyl phenyl ethan-1-one, 4-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethyl acetophenone, 2-bromo-1-4-trifluoromethyl phenyl ethanone, 2-bromo-4'-trifluoromethyl acetophenon, 4-trifluoromethylphenacyl bromide, ethanone, 2-bromo-1-4-trifluoromethyl phenyl, 4'-trifluoromethyl phenacyl bromide, 2-bromo-4'-trifluoromethylacetophenone
FC(F)(F)C1=CC=C(C=C1)C(=O)CBr
267.05
267.04
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Product Identifier
  • 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : 2-Bromo-1-[4-(trifluoromethyl)phenyl]ethan-1-one

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.