missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-1-[5-(2-pyridinyl)-2-thienyl]-1-ethanone, 90%, Thermo Scientific™ Product Code.: 10513002

2-Bromo-1-[5-(2-pyridinyl)-2-thienyl]-1-ethanone, 90%, Thermo Scientific™

Product Code. 10513002
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10513002

Brand: Thermo Scientific Maybridge CC02205DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 306935-06-8
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C11H8BrNOS
MDL Number MFCD02677697
Quantity 1 g
Synonym 2-bromo-1-5-2-pyridinyl-2-thienyl-1-ethanone, 2-bromo-1-5-pyridin-2-yl thiophen-2-yl ethanone, 2-bromo-1-5-pyridin-2-ylthien-2-yl ethan-1-one, 2-bromo-1-5-pyridin-2-yl-thiophen-2-yl-ethanone, 2-bromo-1-5-pyridin-2-yl-2-thienyl ethanone, 2-bromo-1-5-pyridin-2-ylthiophen-2-yl ethanone, 2-bromanyl-1-5-pyridin-2-ylthiophen-2-yl ethanone, ethanone, 2-bromo-1-5-2-pyridinyl-2-thienyl, 2-bromo-1-5-pyridin-2-yl thiophen-2-yl ethan-1-one
InChI Key MJDNWZQQRFCXRU-UHFFFAOYSA-N
SMILES C1=CC=NC(=C1)C2=CC=C(S2)C(=O)CBr
IUPAC Name 2-bromo-1-(5-pyridin-2-ylthiophen-2-yl)ethanone
Molecular Weight (g/mol) 282.155
PubChem CID 2776201
Formula Weight 282.16
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-[5-(2-pyridinyl)-2-thienyl]-1-ethanone
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-1-[5-(2-pyridinyl)-2-thienyl]-1-ethanone, 90%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.