missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -20.0°C |
Density | 1.1800g/mL |
Boiling Point | 72.0°C to 74.0°C |
Flash Point | 18°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3)3CBr |
Refractive Index | 1.4269 to 1.4289 |
Quantity | 100 mL |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0462631 | Certificate of Analysis | 17/05/2024 | 159260025,159261000,159265000 |
A0452526 | Certificate of Analysis | 03/05/2023 | 159260025,159261000,159265000 |
A0407365 | Certificate of Analysis | 13/12/2022 | 159260025,159261000,159265000 |
A0409723 | Certificate of Analysis | 13/12/2022 | 159260025,159261000,159265000 |
A0416681 | Certificate of Analysis | 12/12/2022 | 159260025,159261000,159265000 |
Safety and Handling


Product Identifier
- 2-Bromo-2-methylpropane
- Danger
- Specific target organ toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H225-Highly flammable liquid and vapour.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Bromo-2-methylpropane, 96%, stabilized, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction