missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-2-nitropropane-1,3-diol, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.
Solubility
Soluble in water(280g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | ∼122°C to 132°C |
pH | 5 to 7 |
Quantity | 100 g |
UN Number | UN3241 |
Beilstein | 1705868 |
Merck Index | 14,1447 |
Solubility Information | Soluble in water(280g/L). |
Formula Weight | 199.99 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromo-2-nitropropane-1,3-diol |
Safety and Handling
Product Identifier
- 2-Bromo-2-nitropropane-1,3-diol
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-Bromo-2-nitro-1,3-propanediol
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'