missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-hydroxyacetophenone, 96%, Thermo Scientific Chemicals

Product Code. 10169969
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-37-4
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.04
MDL Number MFCD05664326
InChI Key IEPSGFQQGKPTPM-UHFFFAOYSA-N
PubChem CID 6917206
IUPAC Name 2-bromo-1-(3-hydroxyphenyl)ethanone
SMILES C1=CC(=CC(=C1)O)C(=O)CBr

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 70°C to 74°C
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)C6H4COCH2Br
Quantity 1 g
Synonym 2-bromo-1-3-hydroxyphenyl ethanone, 2-bromo-3'-hydroxyacetophenone, 2-bromo-3-hydroxyacetophenone, ethanone, 2-bromo-1-3-hydroxyphenyl, 3-hydroxyphenacyl bromide, 2-bromo-1-3-hydroxyphenyl ethan-1-one, ethanone,2-bromo-1-3-hydroxyphenyl, 2-bromo-3`-hydroxyacetophenone, 2-bromo-3'-hydroxy-acetophenone, tos-bb-1139
Formula Weight 215.04
Percent Purity 96%
Chemical Name or Material 2-Bromo-3′-hydroxyacetophenone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Bromo-3'-hydroxyacetophenone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.