missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-3'-(trifluoromethyl)acetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11325145
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2003-10-3
Molecular Formula C9H6BrF3O
Molecular Weight (g/mol) 267.05
MDL Number MFCD03094283
InChI Key TZIYNLSEBAYCBZ-UHFFFAOYSA-N

Specifications

Melting Point 20°C to 23°C
Boiling Point 69°C to 72°C
Quantity 1 g
Synonym decoyinine, decoyinin, angustmycin a, antibiotic a 14, decoyinine u-7984, 9-6-deoxy-beta-d-erythro-hex-5-en-2-ulofuranosyl-9h-purin-6-amine, 9-6-deoxy-beta-d-erythro-hex-5-enulofuranosyl adenine, adenine, 9-6-deoxy-beta-d-erythro-hex-5-enulofuranosyl, 9-tetrahydro-3,4-dihydroxy-2-hydroxymethyl-5-methylene-2-furyl adenine, 9h-purin-6-amine, 9-6-deoxy-beta-d-erythro-hex-5-en-2-ulofuranosyl
SMILES FC(F)(F)C1=CC=CC(=C1)C(=O)CBr
IUPAC Name (2R,3R,4S)-2-(6-aminopurin-9-yl)-2-(hydroxymethyl)-5-methylideneoxolane-3,4-diol
PubChem CID 121578
Formula Weight 267.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-3'-(trifluoromethyl)acetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'