Learn More
2-Bromo-4'-chloroacetophenone, 98%, Thermo Scientific Chemicals
47.34 € - 463.31 €
Chemical Identifiers
CAS | 536-38-9 |
---|---|
Molecular Formula | C8H6BrClO |
Molecular Weight (g/mol) | 233.489 |
MDL Number | MFCD00000625 |
InChI Key | FLAYZKKEOIAALB-UHFFFAOYSA-N |
Synonym | 2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone |
PubChem CID | 68303 |
IUPAC Name | 2-bromo-1-(4-chlorophenyl)ethanone |
SMILES | C1=CC(=CC=C1C(=O)CBr)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424344
|
Thermo Scientific Alfa Aesar
A12607.14 |
25 g |
47.34 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434344
|
Thermo Scientific Alfa Aesar
A12607.22 |
100 g |
135.68 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444344
|
Thermo Scientific Alfa Aesar
A12607.36 |
500 g |
463.31 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Bromo-4'-chloroacetophenone is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-4′-chloroacetophenone is used as pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Chemical Identifiers
536-38-9 | |
233.489 | |
FLAYZKKEOIAALB-UHFFFAOYSA-N | |
68303 | |
C1=CC(=CC=C1C(=O)CBr)Cl |
C8H6BrClO | |
MFCD00000625 | |
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone | |
2-bromo-1-(4-chlorophenyl)ethanone |
Specifications
536-38-9 | |
C8H6BrClO | |
UN3261 | |
14,2153 | |
Insoluble in water. | |
C1=CC(=CC=C1C(=O)CBr)Cl | |
233.489 | |
233.5 | |
2-Bromo-4'-chloroacetophenone |
94°C to 98°C | |
MFCD00000625 | |
607603 | |
2-bromo-4'-chloroacetophenone, 2-bromo-1-4-chlorophenyl ethanone, 4-chlorophenacyl bromide, p-chlorophenacyl bromide, 2-bromo-1-4-chlorophenyl ethan-1-one, ethanone, 2-bromo-1-4-chlorophenyl, 4'-chlorophenacyl bromide, 2-bromo-p-chloroacetophenone, p-bromoacetyl chlorobenzene, 4'-chloro-2-bromoacetophenone | |
FLAYZKKEOIAALB-UHFFFAOYSA-N | |
2-bromo-1-(4-chlorophenyl)ethanone | |
68303 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 208-631-5
missing translation for 'rtecsNumber' : AM5978800
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.