missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-4-fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals Product Code.: 11326639

2-Bromo-4-fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Product Code. 11326639
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11326639

Brand: Thermo Scientific Alfa Aesar H26211.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193090-60-7
Molecular Formula C7H4BrF4N
Molecular Weight (g/mol) 258.01
MDL Number MFCD04973755
InChI Key ZRMRCSMDONHRKT-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-5-trifluoromethyl aniline, 5-amino-4-bromo-2-fluorobenzotrifluoride, benzenamine, 2-bromo-4-fluoro-5-trifluoromethyl, 2-bromo-4-fluoro-5'-trifluoromethyl aniline, 2-bromo-4-fluoro-5-trifluoromethyl-phenylamine, 2-bromo-4-fluoro-5-trifluoromethyl-benzenamine, acmc-209ewq, ksc496o8r, 2-bromo-4-fluoro-5-trifluoromethyl benzenamine, 2-bromo-4-fluoro-5-trifluoromethyl phenylamine
PubChem CID 24721365
IUPAC Name 2-bromo-4-fluoro-5-(trifluoromethyl)aniline
SMILES NC1=C(Br)C=C(F)C(=C1)C(F)(F)F

Specifications

Melting Point 24°C to 29°C
Quantity 5 g
UN Number UN2811
Formula Weight 258.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-4-fluoro-5-(trifluoromethyl)aniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-4-fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.