missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4'-(trifluoromethoxy)acetophenone, 97%, Thermo Scientific Chemicals

28.20 € - 248.24 €

Chemical Identifiers

CAS 103962-10-3
Molecular Formula C9H6BrF3O2
Molecular Weight (g/mol) 283.044
MDL Number MFCD00052333
InChI Key AOAGGWLQIILIIV-UHFFFAOYSA-N
Synonym 4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
PubChem CID 2778691
IUPAC Name 2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15428706
SDS View Documents
Thermo Scientific Alfa Aesar
H64105.MD
250 mg N/A
N/A
15411159
SDS View Documents
Thermo Scientific Alfa Aesar
H64105.03
1 g N/A
N/A
15421159
SDS View Documents
Thermo Scientific Alfa Aesar
H64105.06
5 g N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

103962-10-3
283.044
AOAGGWLQIILIIV-UHFFFAOYSA-N
2778691
C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F
C9H6BrF3O2
MFCD00052333
4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone

Specifications

103962-10-3
Yellow
C9H6BrF3O2
7479010
AOAGGWLQIILIIV-UHFFFAOYSA-N
2-bromo-1-[4-(trifluoromethoxy)phenyl]ethanone
2778691
97%
2-Bromo-4'-(trifluoromethoxy)acetophenone
48°C to 55°C
>110°C (230°F)
MFCD00052333
4-trifluoromethoxy phenacyl bromide, 2-bromo-1-4-trifluoromethoxy phenyl ethanone, 2-bromo-4'-trifluoromethoxy acetophenone, 2-bromo-1-4-trifluoromethoxy phenyl ethan-1-one, 2-bromo-4'-trifluoromethoxyacetophenone, 2-bromo-1-4-trifluoromethoxy-phenyl-ethanone, p-trifluoromethoxy phenacyl bromide, 1-4-trifluoromethoxyphenyl-2-bromoethan-1-one, ethanone,2-bromo-1-4-trifluoromethoxy phenyl
C1=CC(=CC=C1C(=O)CBr)OC(F)(F)F
283.044
283.04
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.