missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific Chemicals Product Code.: 11421330

2-Bromo-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific Chemicals

Product Code. 11421330
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11421330

Brand: Thermo Scientific Alfa Aesar H26389.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 205371-26-2
Molecular Formula C7H4BrF3O2
Molecular Weight (g/mol) 257.01
MDL Number MFCD06660162
InChI Key RHRRKORKKIVAGJ-UHFFFAOYSA-N
Synonym 2-bromo-5-trifluoromethoxy phenol, phenol, 2-bromo-5-trifluoromethoxy, 2-hydroxy-4-trifluoromethoxy bromobenzene, 1-bromo-2-hydroxy-4-trifluoromethoxy benzene, pubchem15610, acmc-209fbt, rhrrkorkkivagj-uhfffaoysa, phenol,2-bromo-5-trifluoromethoxy, 2-bromanyl-5-trifluoromethyloxy phenol
PubChem CID 17750742
IUPAC Name 2-bromo-5-(trifluoromethoxy)phenol
SMILES OC1=C(Br)C=CC(OC(F)(F)F)=C1

Specifications

Refractive Index 1.4815
Quantity 5 g
Formula Weight 257.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethoxy)phenol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10182946 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26389.14,H26389.06
Lot Number10146511 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26389.14,H26389.06
Lot NumberK13S037 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26389.14,H26389.06
Lot Number10141151 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26389.14,H26389.06
Lot Number10181609 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26389.14,H26389.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.