missing translation for 'onlineSavingsMsg'
Learn More

2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole, Thermo Scientific™

Product Code. p-4391234
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10595270

Brand: Thermo Scientific Maybridge CC72310DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 37461-61-3
Molecular Formula C3BrF3N2S
Molecular Weight (g/mol) 233.01
MDL Number MFCD00015535
InChI Key LGTVLLPQCMJOGT-UHFFFAOYSA-N
PubChem CID 12874685
IUPAC Name 2-bromo-5-(trifluoromethyl)-1,3,4-thiadiazole
SMILES FC(F)(F)C1=NN=C(Br)S1

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-bromo-5-trifluoromethyl-1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4 thiadiazole, 2-bromo-5-trifluoromethyl-1,3,4-thiadiazol, 5-bromo-2-trifluoromethyl-1,3,4-thiadiazole, 1,3,4-thiadiazole,2-bromo-5-trifluoromethyl, 1,3,4-thiadiazole, 2-bromo-5-trifluoromethyl, 2-bromanyl-5-trifluoromethyl-1,3,4-thiadiazole
Formula Weight 233.01
Percent Purity 97%
Chemical Name or Material 2-bromo-5-(trifluoromethyl)-1, 3, 4-thiadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.