missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-6-chloro-4-fluorophenol, 97%, Thermo Scientific™

Product Code. 11342719
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886499-83-8
Molecular Formula C6H3BrClFO
Molecular Weight (g/mol) 225.44
MDL Number MFCD04115954
InChI Key CRWILTIFGXJONJ-UHFFFAOYSA-N
PubChem CID 3818635
IUPAC Name 2-bromo-6-chloro-4-fluorophenol
SMILES OC1=C(Cl)C=C(F)C=C1Br

Specifications

Melting Point 38°C to 40°C
Quantity 1 g
Formula Weight 225.45
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-chloro-4-fluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'