Learn More
2-Bromo-6-pyrrolidin-1-ylpyridine, ≥97%, Thermo Scientific™
16.10 € - 177.00 €
Chemical Identifiers
CAS | 230618-41-4 |
---|---|
Molecular Formula | C9H11BrN2 |
Molecular Weight (g/mol) | 227.105 |
MDL Number | MFCD08271892 |
InChI Key | HZOWJDJNPISFJC-UHFFFAOYSA-N |
Synonym | 2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl |
PubChem CID | 15470277 |
IUPAC Name | 2-bromo-6-pyrrolidin-1-ylpyridine |
SMILES | C1CCN(C1)C2=NC(=CC=C2)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10010213
|
Thermo Scientific Maybridge
CC38010CB |
250 mg |
16.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10706463
|
Thermo Scientific Maybridge
CC38010DA |
1 g |
46.70 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10312305
|
Thermo Scientific Maybridge
CC38010DE |
5 g |
177.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
230618-41-4 | |
227.105 | |
HZOWJDJNPISFJC-UHFFFAOYSA-N | |
15470277 | |
C1CCN(C1)C2=NC(=CC=C2)Br |
C9H11BrN2 | |
MFCD08271892 | |
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl | |
2-bromo-6-pyrrolidin-1-ylpyridine |
Specifications
230618-41-4 | |
100.0 | |
Amber Glass Bottle | |
MFCD08271892 | |
HZOWJDJNPISFJC-UHFFFAOYSA-N | |
2-bromo-6-pyrrolidin-1-ylpyridine | |
15470277 | |
90% |
89.5 | |
90% | |
C9H11BrN2 | |
2-bromo-6-pyrrolidin-1-yl pyridine, 2-bromo-6-pyrrolidin-1-yl-pyridine, acmc-209g0q, 2-bromo-6-pyrrolizinopyridine, 6-bromo-2-pyrrolidinylpyridine, 2-bromo-6-1-pyrrolidinyl pyridine, 2-bromanyl-6-pyrrolidin-1-yl-pyridine, 2-bromo-6-pyrrolidine-1-yl pyridine, pyridine,2-bromo-6-1-pyrrolidinyl | |
C1CCN(C1)C2=NC(=CC=C2)Br | |
227.105 | |
227.1 | |
2-Bromo-6-pyrrolidin-1-ylpyridine |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.