Learn More
2-Bromobenzeneboronic acid, 98%, Thermo Scientific Chemicals
43.11 € - 109.55 €
Chemical Identifiers
CAS | 244205-40-1 |
---|---|
Molecular Formula | C6H6BBrO2 |
Molecular Weight (g/mol) | 200.83 |
MDL Number | MFCD01114672 |
InChI Key | PLVCYMZAEQRYHJ-UHFFFAOYSA-N |
Synonym | 2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acidShow More |
PubChem CID | 2773294 |
IUPAC Name | (2-bromophenyl)boronic acid |
SMILES | OB(O)C1=CC=CC=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319425
|
Thermo Scientific Alfa Aesar
L18581.03 |
1 g |
43.11 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11329425
|
Thermo Scientific Alfa Aesar
L18581.06 |
5 g |
109.55 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromobenzeneboronic acid is used to promote greener amidations of carboxylic acids and amines in catalytic amounts. This technology avoids the requirement of preactivation of the carboxylic acid or use of coupling reagents.
Solubility
Soluble in methanol. Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
244205-40-1 | |
200.83 | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
2773294 | |
OB(O)C1=CC=CC=C1Br |
C6H6BBrO2 | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acidShow More | |
(2-bromophenyl)boronic acid |
Specifications
244205-40-1 | |
C6H6BBrO2 | |
8542615 | |
Soluble in methanol. Slightly soluble in water. | |
OB(O)C1=CC=CC=C1Br | |
200.83 | |
200.83 | |
2-Bromobenzeneboronic acid |
108°C to 110°C | |
MFCD01114672 | |
2-bromophenyl boronic acid, o-bromophenylboronic acid, 2-bromobenzeneboronic acid, 2-bromophenylboronicacid, boronic acid, 2-bromophenyl, bromobenzeneboronic acid, pubchem13095, 2-bromo phenyl boronic acid, 2-bromo-phenyl boronic acid | |
PLVCYMZAEQRYHJ-UHFFFAOYSA-N | |
(2-bromophenyl)boronic acid | |
2773294 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.