missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoethyl acetate, 97%, Thermo Scientific Chemicals

Product Code. 10493340
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-68-4
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00000235
InChI Key RGHQKFQZGLKBCF-UHFFFAOYSA-N
Synonym bromoethyl acetate, 1-acetoxy-2-bromoethane, 2-bromoethylacetate, ethanol, 2-bromo-, acetate, 1-bromo-2-acetoxyethane, acetic acid 2-bromoethyl ester, unii-nrz256o7qb, ethanol, 2-bromo-, 1-acetate, beta-bromoethyl acetate, nrz256o7qb
PubChem CID 13564
IUPAC Name 2-bromoethyl acetate
SMILES CC(=O)OCCBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -13.8°C
Color Colorless to Yellow
Density 1.5100g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2CH2Br
Refractive Index 1.4547
Quantity 100 g
Specific Gravity 1.51
Formula Weight 167
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Bromoethyl acetate
Show More Show Less
compliance-icons
Produktidentifikator
  • 2-Bromoethyl acetate
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'