missing translation for 'onlineSavingsMsg'
Learn More
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals Product Code.: 10427290

2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals

Product Code. 10427290 Shop All Thermo Scientific Products
50 mL, Glass bottle
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10427290

Brand: Thermo Scientific Acros 106990500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-85-1
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000282
InChI Key IPWBFGUBXWMIPR-UHFFFAOYSA-N
Synonym 2-bromofluorobenzene, o-bromofluorobenzene, 2-fluorobromobenzene, o-fluorobromobenzene, 2-bromfluorbenzen, benzene, 1-bromo-2-fluoro, 1-fluoro-2-bromobenzene, benzene, 1-fluoro-2-bromo, ofbb, benzene, bromofluoroShow MoreShow Less
PubChem CID 61259
IUPAC Name 1-bromo-2-fluorobenzene
SMILES C1=CC=C(C(=C1)F)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6000g/mL
Boiling Point 155.0°C to 157.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H4F
Refractive Index 1.5327 to 1.5347
Quantity 50 mL
Fieser 01,77
Specific Gravity 1.6
Solubility Information Solubility in water: insoluble in water
Formula Weight 175
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Bromofluorobenzene, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453103 Certificate TypeCertificate of Analysis Date16/05/2023 Catalog Number106990100,106990500
Lot NumberA0417991 Certificate TypeCertificate of Analysis Date26/09/2022 Catalog Number106990100,106990500
Lot NumberA0405601 Certificate TypeCertificate of Analysis Date23/08/2021 Catalog Number106990100,106990500
Lot NumberA0397953 Certificate TypeCertificate of Analysis Date24/12/2020 Catalog Number106990100,106990500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromofluorobenzene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.