missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-1,3-benzothiazole, 95%, Thermo Scientific™

Product Code. p-4509937 Shop All Thermo Scientific Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10571642

Brand: Thermo Scientific CC06008CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 106086-78-6
Molecular Formula C8H6BrNS
Molecular Weight (g/mol) 228.107
InChI Key WFLCAOGKZQTOIG-UHFFFAOYSA-N
Synonym 2-bromomethyl-1,3-benzothiazole, 2-bromomethyl benzo d thiazole, 2-bromomethyl benzothiazole, benzothiazole, 2-bromomethyl, benzo d thiazol-2-yl methyl bromide, zlchem 58, pubchem3912, acmc-1bpju, 2-bromomethylbenzothiazole
PubChem CID 2776258
IUPAC Name 2-(bromomethyl)-1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=C(S2)CBr

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 228.11
Percent Purity 95%
Chemical Name or Material 2-(Bromomethyl)-1, 3-benzothiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.