missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide, 97%, Thermo Scientific™

Product Code. 10626994
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 934570-40-8
Molecular Formula C9H10Br2N2
Molecular Weight (g/mol) 306.001
MDL Number MFCD09702343
InChI Key WHMCWOHHFOGLGP-UHFFFAOYSA-N
Synonym 2-bromomethyl-1-methyl-1h-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzo d imidazole hydrobromide, 2-bromomethyl-1-methyl-1,3-benzodiazole hydrobromide, 2-bromomethyl-1-methyl-benzimidazole hydrobromide, 2-bromomethyl-1-methyl-1h-benzimidazole hbr, 2-bromomethyl-1-methyl-1h-benzimidazole-hydrogen bromide 1/1
PubChem CID 24229451
IUPAC Name 2-(bromomethyl)-1-methylbenzimidazole;hydrobromide
SMILES CN1C2=CC=CC=C2N=C1CBr.Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 306
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-1-methyl-1H-benzimidazole hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.