missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)-1-(phenylsulfonyl)-1H-indole, 97%, Thermo Scientific™

Product Code. 10170722
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 337508-54-0
Molecular Formula C15H12BrNO2S
Molecular Weight (g/mol) 350.23
MDL Number MFCD03086092
InChI Key SYYPBRKVYRCLJS-UHFFFAOYSA-N
Synonym 2-bromomethyl-1-phenylsulfonyl-1h-indole, 1-benzenesulfonyl-2-bromomethyl indole, 1h-indole,2-bromomethyl-1-phenylsulfonyl
PubChem CID 2776225
IUPAC Name 1-(benzenesulfonyl)-2-(bromomethyl)indole
SMILES BrCC1=CC2=CC=CC=C2N1S(=O)(=O)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 350.23
Percent Purity 97%
Chemical Name or Material 2-(Bromomethyl)-1-(phenylsulfonyl)-1H-indole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'