missing translation for 'onlineSavingsMsg'
Learn More

2-(Bromomethyl)naphthalene, 96%, Thermo Scientific Chemicals

Product Code. 11490408
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water (reacts), and chloroform.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 939-26-4
Molecular Formula C11H9Br
Molecular Weight (g/mol) 221.10
MDL Number MFCD00004123
InChI Key RUHJZSZTSCSTCC-UHFFFAOYSA-N
Synonym 2-bromomethyl naphthalene, naphthalene, 2-bromomethyl, 2-naphthylmethyl bromide, 2-naphthyl bromomethane, 2-bromomethyl napthalene, .beta.-bromomethyl naphthalene, 2-brommethyl naphthalen, zlchem 566, 2-menaphthyl bromide
PubChem CID 70320
IUPAC Name 2-(bromomethyl)naphthalene
SMILES BrCC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 51°C to 55°C
Boiling Point 213°C (100 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 636546
Solubility Information Soluble in water (reacts),and chloroform.
Formula Weight 221.1
Percent Purity 96%
Chemical Name or Material 2-(Bromomethyl)naphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.