Learn More
2-(Bromomethyl)pyridine hydrobromide, 98%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 149.0°C to 152.0°C |
Color | Brown to White |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Glass bottle |
Linear Formula | BrCH2(C5H3N) |
Quantity | 25 g |
Formula Weight | 252.94 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0458688 | Certificate of Analysis | 31/01/2025 | 370110050,370110250,370111000 |
A0438766 | Certificate of Analysis | 15/02/2024 | 370110050,370110250,370111000 |
A0458688 | Certificate of Analysis | 04/01/2024 | 370110050,370110250,370111000 |
A0386480 | Certificate of Analysis | 28/02/2022 | 370110050,370110250,370111000 |
A0438766 | Certificate of Analysis | 25/02/2022 |
Safety and Handling


- 2-(Bromomethyl)pyridine hydrobromide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 2-(Bromomethyl)pyridine hydrobromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.